Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-1-cyclopentene-1-carbonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A183484-100G A183484-1G A183484-25G A183484-5G
ALA183484-100GEA 460.66 USD InStock
ALA183484-100G ALA183484-1G ALA183484-25G ALA183484-5G
2-Amino-1-cyclopentene-1-carbonitrile ≥98%
2-Amino-1-cyclopentene-1-carbonitrile


Formula: C₆H₈N₂
MW: 108.144 g/mol
Melting Pt: 147-148°C
Storage Temperature: Refrigerator
MDL Number: MFCD00517551
CAS Number: 2941-23-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty