Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-(methoxycarbonyl)benzeneboronic acid hydrochloride ≥95% (contains varying amounts of Anhydride)

Supplier: ALADDIN SCIENTIFIC
Warning

A184145-1G A184145-250MG A184145-5G
ALA184145-1GEA 220.31 USD InStock
ALA184145-1G ALA184145250MG ALA184145-5G
2-Amino-4-(methoxycarbonyl)benzeneboronic acid hydrochloride ≥95% (contains varying amounts of Anhydride)
2-Amino-4-(methoxycarbonyl)benzeneboronic acid hydrochloride


Formula: C₈H₁₀BNO₄·HCl
MW: 231.44 g/mol
Melting Pt: 200-204°C
Storage Temperature: Ambient
MDL Number: MFCD02258941
CAS Number: 380430-55-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty