Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Aminomethyl)tetrahydropyran hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A184237-100G A184237-25G A184237-5G
ALA184237-100GEA 805.51 USD InStock
ALA184237-100G ALA184237-25G ALA184237-5G
4-(Aminomethyl)tetrahydropyran hydrochloride ≥98%
4-(Aminomethyl)tetrahydropyran hydrochloride


Formula: C₆H₁₄ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD03411928
CAS Number: 389621-78-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty