Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenyl-2-sulfoacetic acid ≥80%

Supplier: ALADDIN SCIENTIFIC
Warning

A184434-100G A184434-1G A184434-25G A184434-5G
ALA184434-100GEA 126.26 USD InStock
ALA184434-100G ALA184434-1G ALA184434-25G ALA184434-5G
2-Phenyl-2-sulfoacetic acid ≥80%
2-Phenyl-2-sulfoacetic acid


Formula: C₈H₈O₅S
Storage Temperature: Ambient
MDL Number: MFCD09952336
CAS Number: 41360-32-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty