Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3alpha-Hydroxy-7-oxo-5beta-cholanic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A184646-25G A184646-5G A184646-1G
ALA184646-25GEA 1429.66 USD InStock
ALA184646-25G ALA184646-5G ALA184646-1G
3alpha-Hydroxy-7-oxo-5beta-cholanic acid ≥98%
3alpha-Hydroxy-7-oxo-5beta-cholanic acid


Formula: C₂₄H₃₈O₄
Storage Temperature: Ambient
MDL Number: MFCD00271393
CAS Number: 4651-67-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty