Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-chloropyridine-3-carboxylic acid ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

A184995-1G A184995-250MG A184995-25G A184995-5G
ALA184995-1GEA 37.91 USD InStock
ALA184995-1G ALA184995250MG ALA184995-25G ALA184995-5G
2-Amino-5-chloropyridine-3-carboxylic acid ≥96%
2-Amino-5-chloropyridine-3-carboxylic acid


Formula: C₆H₅ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09839254
CAS Number: 52833-93-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty