Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

H-Phe(4-NH2)-OH·HCl ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

A185605-1G A185605-250MG A185605-5G
ALA185605-1GEA 28.41 USD InStock
ALA185605-1G ALA185605250MG ALA185605-5G
H-Phe(4-NH2)-OH·HCl ≥96%
H-Phe(4-NH2)-OH·HCl


Formula: C₉H₁₂N₂O₂·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00012986
CAS Number: 62040-55-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty