Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-fluorobenzamide ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A185682-5G A185682-1G A185682-250MG A185682-25G
ALA185682-5GEA 158.56 USD InStock
ALA185682-5G ALA185682-1G ALA185682250MG ALA185682-25G
2-Amino-5-fluorobenzamide ≥97%
2-Amino-5-fluorobenzamide


Formula: C₇H₇FN₂O
Melting Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD03428522
CAS Number: 63069-49-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty