Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-3-chlorophenylboronic acid pinacol ester ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A186244-1G A186244-250MG A186244-5G
ALA186244-1GEA 80.66 USD InStock
ALA186244-1G ALA186244250MG ALA186244-5G
4-Amino-3-chlorophenylboronic acid pinacol ester ≥98%
4-Amino-3-chlorophenylboronic acid pinacol ester


Formula: C₁₂H₁₇BClNO₂
Storage Temperature: Ambient
MDL Number: MFCD09998359
CAS Number: 721960-43-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty