Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-4-chlorophenylboronic acid pinacol ester ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A187011-1G A187011-250MG A187011-5G
ALA187011-1GEA 42.66 USD InStock
ALA187011-1G ALA187011250MG ALA187011-5G
3-Amino-4-chlorophenylboronic acid pinacol ester ≥98%
3-Amino-4-chlorophenylboronic acid pinacol ester


Formula: C₁₂H₁₇BClNO₂
Melting Pt: 142-144°C
Storage Temperature: Refrigerator
MDL Number: MFCD06659926
CAS Number: 850567-56-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty