Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-(Allylcarbamoyl)phenyl)boronic acid ≥97%

Supplier: ALADDIN SCIENTIFIC

A187031-1G A187031-250MG A187031-5G
ALA187031-1GEA 110.11 USD InStock
ALA187031-1G ALA187031250MG ALA187031-5G
(4-(Allylcarbamoyl)phenyl)boronic acid ≥97%
(4-(Allylcarbamoyl)phenyl)boronic acid


Formula: C₁₀H₁₂BNO₃
Storage Temperature: Ambient
MDL Number: MFCD04115674
CAS Number: 850568-20-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty