Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-6-bromo-2-methoxypyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A187873-5G A187873-10G A187873-1G A187873-250MG A187873-25G
ALA187873-5GEA 87.31 USD InStock
ALA187873-5G ALA187873-10G ALA187873-1G ALA187873250MG ALA187873-25G
3-Amino-6-bromo-2-methoxypyridine ≥97%
3-Amino-6-bromo-2-methoxypyridine


Formula: C₆H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD06738312
CAS Number: 89466-18-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty