Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(5-Amino-1,2,4-thiadiazol-3-yl)-2-(methoxyimino)acetic acid ≥98%

Supplier: ALADDIN SCIENTIFIC

A189191-25G A189191-5G A189191-1G
ALA189191-25GEA 412.21 USD InStock
ALA189191-25G ALA189191-5G ALA189191-1G
2-(5-Amino-1,2,4-thiadiazol-3-yl)-2-(methoxyimino)acetic acid ≥98%
2-(5-Amino-1,2,4-thiadiazol-3-yl)-2-(methoxyimino)acetic acid


Formula: C₅H₆N₄O₃S
Storage Temperature: Refrigerator
MDL Number: MFCD08444061
CAS Number: 72217-12-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty