Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3,5-difluorobenzonitrile ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A190410-1G A190410-250MG A190410-5G
ALA190410-1GEA 27.46 USD InStock
ALA190410-1G ALA190410250MG ALA190410-5G
2-Amino-3,5-difluorobenzonitrile ≥97%
2-Amino-3,5-difluorobenzonitrile


Formula: C₇H₄F₂N₂
MDL Number: MFCD18826239
CAS Number: 126674-94-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty