Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(1,1,1,5,5,5-Hexamethyl-3-((trimethylsilyl)oxy)trisiloxan-3-yl)propyl acrylate ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

A191490-1G A191490-25G A191490-5G
ALA191490-1GEA 9.41 USD InStock
ALA191490-1G ALA191490-25G ALA191490-5G
3-(1,1,1,5,5,5-Hexamethyl-3-((trimethylsilyl)oxy)trisiloxan-3-yl)propyl acrylate ≥96%
3-(1,1,1,5,5,5-Hexamethyl-3-((trimethylsilyl)oxy)trisiloxan-3-yl)propyl acrylate


Formula: C₁₅H₃₆O₅Si₄
MW: 408.79 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00054802
CAS Number: 17096-12-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty