Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminopentanoic acid ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A191758-100MG A191758-1G A191758-250MG
ALA191758100MGEA 28.41 USD InStock
ALA191758100MG ALA191758-1G ALA191758250MG
3-Aminopentanoic acid ≥95%
3-Aminopentanoic acid


Formula: C5H11NO2
Storage Temperature: Refrigerator
CAS Number: 18664-78-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty