Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Methoxyphenyl)glycine ≥95%

Supplier: ALADDIN SCIENTIFIC

A192458-10G A192458-250MG A192458-50MG A192458-5G
ALA192458-10GEA 647.81 USD InStock
ALA192458-10G ALA192458250MG ALA192458-50MG ALA192458-5G
2-(2-Methoxyphenyl)glycine ≥95%
2-(2-Methoxyphenyl)glycine


Formula: C₉H₁₁NO₃
MDL Number: MFCD02662408
CAS Number: 271583-17-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty