Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-3-(4-bromophenyl)propionic acid ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A193198-1G A193198-250MG A193198-25G A193198-5G
ALA193198-1GEA 16.06 USD InStock
ALA193198-1G ALA193198250MG ALA193198-25G ALA193198-5G
3-Amino-3-(4-bromophenyl)propionic acid ≥95%
3-Amino-3-(4-bromophenyl)propionic acid


Formula: C₉H₁₀BrNO₂
MW: 244.0900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00187112
CAS Number: 39773-47-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty