Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-5-(trifluoromethyl)aniline ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A193474-1G A193474-250MG A193474-25G A193474-5G
ALA193474-1GEA 51.21 USD InStock
ALA193474-1G ALA193474250MG ALA193474-25G ALA193474-5G
2-Hydroxy-5-(trifluoromethyl)aniline ≥98%
2-Hydroxy-5-(trifluoromethyl)aniline


Formula: C₇H₆F₃NO
MW: 177.1200 g/mol
Boiling Pt: 234.5°C
Storage Temperature: Ambient
MDL Number: MFCD01731750
CAS Number: 454-81-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty