Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-chlorophenol ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A193993-1G A193993-250MG A193993-50MG
ALA193993-1GEA 116.76 USD InStock
ALA193993-1G ALA193993250MG ALA193993-50MG
2-Amino-3-chlorophenol ≥98%
2-Amino-3-chlorophenol


Formula: C₆H₆ClNO
MDL Number: MFCD11848418
CAS Number: 56962-00-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty