Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Acetoxyethoxy)methyl Acetate ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A194100-100G A194100-1G A194100-25G A194100-5G
ALA194100-100GEA 1608.26 USD InStock
ALA194100-100G ALA194100-1G ALA194100-25G ALA194100-5G
(2-Acetoxyethoxy)methyl Acetate ≥97%
(2-Acetoxyethoxy)methyl Acetate


Formula: C₇H₁₂O₅
Boiling Pt: 116 °C
Density: 1.14 g/cm³
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09031327
CAS Number: 59278-00-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty