Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6-chlorobenzonitrile ≥96%

Supplier: ALADDIN SCIENTIFIC
Danger

A194436-1G A194436-25G A194436-5G
ALA194436-1GEA 21.76 USD InStock
ALA194436-1G ALA194436-25G ALA194436-5G
2-Amino-6-chlorobenzonitrile ≥96%
2-Amino-6-chlorobenzonitrile


Formula: C₇H₅ClN₂
MW: 152.58
Boiling Pt: 301
Melting Pt: 132-138°C
Storage Temperature: Refrigerator
MDL Number: MFCD00015546
CAS Number: 6575-11-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty