Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-N-(9H-xanthen-9-yl)benzenesulfonamide ≥98%

Supplier: ALADDIN SCIENTIFIC

A274890-100MG A274890-10MG A274890-200MG A274890-25MG A274890-2MG A274890-50MG A274890-5MG
ALA274890100MGEA 564.21 USD InStock
ALA274890100MG ALA274890-10MG ALA274890200MG ALA274890-25MG ALA274890-2MG ALA274890-50MG ALA274890-5MG
4-Methyl-N-(9H-xanthen-9-yl)benzenesulfonamide ≥98%
4-Methyl-N-(9H-xanthen-9-yl)benzenesulfonamide
Store at Room Temperature. The product can be stored for up to 12 months.


Formula: C₂₀H₁₇NO₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00379273
CAS Number: 6326-06-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty