Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Astragaloside A ≥98%

Supplier: ALADDIN SCIENTIFIC

A274907-1G A274907-250MG A274907-25G A274907-5G
ALA274907-1GEA 98.71 USD InStock
ALA274907-1G ALA274907250MG ALA274907-25G ALA274907-5G
Astragaloside A ≥98%
Astragaloside A
Astragaloside A (Astragaloside IV) is the primary pure saponin isolated from Astragalus membranaceus, which has been widely used for the therapy of cardiovascular diseases.


Formula: C₄₁H₆₈O₁₄
MW: 784.970 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00210476
CAS Number: 83207-58-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty