Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aminopurvalanol A ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC

A274969-25MG A274969-50MG A274969-5MG A274969-100MG A274969-10MG A274969-1MG
ALA274969-25MGEA 366.61 USD InStock
ALA274969-25MG ALA274969-50MG ALA274969-5MG ALA274969100MG ALA274969-10MG ALA274969-1MG
Aminopurvalanol A ≥99%, Moligand™
Aminopurvalanol A
Product Introduction:Aminopurvalanol A is a potent, selective, and cell permeable inhibitor of Cyclins/Cdk complexes. Aminopurvalanol A preferentially targets the G2/M-phase transition inhibiting cancer cell differentiation. Aminopurvalanol A causes the inhibition of sperm fertilizing ability via the inhibition of physiological capacitation-dependent actin polymerization.


Formula: C₁₉H₂₆ClN₇O
MW: 403.9090 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798384
CAS Number: 220792-57-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty