Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tyrphostin B46 ≥98%

Supplier: ALADDIN SCIENTIFIC

A275199-100MG A275199-10MG A275199-250MG A275199-25MG A275199-50MG
ALA275199100MGEA 376.11 USD InStock
ALA275199100MG ALA275199-10MG ALA275199250MG ALA275199-25MG ALA275199-50MG
Tyrphostin B46 ≥98%
Tyrphostin B46
Store at +4°C. The product can be stored for up to 12 months.


Formula: C₁₉H₁₈N₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00209865
CAS Number: 133550-34-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty