Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Arachidonylglycine ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC

A275341-100MG A275341-25MG A275341-5MG
ALA275341100MGEA 831.16 USD InStock
ALA275341100MG ALA275341-25MG ALA275341-5MG
N-Arachidonylglycine ≥98%, Moligand™
N-Arachidonylglycine
Product Describtion:N-arachidonoylglycine (NAgly) is an endogenous ligand and a signaling lipid. It belongs to the eicosanoid family.NAgly is a natural ligand to activate G-protein coupled receptor, namely GPR18.It is found in rat brain and spinal cord.


Formula: C₂₂H₃₅NO₃
MW: 361.526 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03791313
CAS Number: 179113-91-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty