Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Arsenic(III) iodide ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A283387-100G A283387-25G A283387-5G
ALA283387-100GEA 1232.06 USD InStock
ALA283387-100G ALA283387-25G ALA283387-5G
Arsenic(III) iodide ≥98%
Arsenic(III) iodide
Product Application:Arsenic(III) iodide is mainly used to treat rheumatism, arthritis, malaria, trypanosome infections, tuberculosis, and diabetes.


Formula: AsI₃
MW: 455.64
Boiling Pt: 400
Melting Pt: 146
Density: 4.39
Storage Temperature: Refrigerator
MDL Number: MFCD00014170
CAS Number: 7784-45-4
UN: 1557
ADR: 6.1 I
Merck Index: 12,00843
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty