Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Antimony(III) bromide ≥99%

Supplier: ALADDIN SCIENTIFIC
Danger

A283455-1G A283455-5G
ALA283455-1GEA 9.41 USD InStock
ALA283455-1G ALA283455-5G
Antimony(III) bromide ≥99%
Antimony(III) bromide
Application:Antimony(III) bromide is used in the synthesis of antimony compounds, in semiconductors and chemical analysis. It acts as a fire retardant in polyethylene polymers, as mordant and in dyeing.


Formula: Br₃Sb
Boiling Pt: 280°C
Melting Pt: 96.6°C
Density: 4.148 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00016317
CAS Number: 7789-61-9
UN: 1549(solid)3141(solution)
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty