Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-2-(7-Fluoro-1-isobutyl-2-oxoindolin-3-ylidene)acetic acid ≥98% (by HPLC), Moligand™

Supplier: ALADDIN SCIENTIFIC

A286598-100MG A286598-10MG A286598-25MG A286598-5MG
ALA286598100MGEA 723.81 USD InStock
ALA286598100MG ALA286598-10MG ALA286598-25MG ALA286598-5MG
(E)-2-(7-Fluoro-1-isobutyl-2-oxoindolin-3-ylidene)acetic acid ≥98% (by HPLC), Moligand™
(E)-2-(7-Fluoro-1-isobutyl-2-oxoindolin-3-ylidene)acetic acid


Formula: C₁₄H₁₄FNO₃
Storage Temperature: Freezer
MDL Number: MFCD29079014
CAS Number: 1190217-35-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty