Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Apilimod mesylate ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A286867-100MG A286867-10MG A286867-25MG A286867-50MG A286867-5MG
ALA286867100MGEA 195.61 USD InStock
ALA286867100MG ALA286867-10MG ALA286867-25MG ALA286867-50MG ALA286867-5MG
Apilimod mesylate ≥98% (by HPLC)
Apilimod mesylate


Formula: C₂₅H₃₄N₆O₈S₂
MW: 610.70 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28167749
CAS Number: 870087-36-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty