Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-6-(((3-Fluoro-4-(trifluoromethoxy)phenyl)(3-fluoropyridin-2-yl)methyl)carbamoyl)nicotinic acid ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A287015-10MG A287015-25MG A287015-50MG A287015-5MG
ALA287015-10MGEA 150.01 USD InStock
ALA287015-10MG ALA287015-25MG ALA287015-50MG ALA287015-5MG
(S)-6-(((3-Fluoro-4-(trifluoromethoxy)phenyl)(3-fluoropyridin-2-yl)methyl)carbamoyl)nicotinic acid ≥98% (by HPLC)
(S)-6-(((3-Fluoro-4-(trifluoromethoxy)phenyl)(3-fluoropyridin-2-yl)methyl)carbamoyl)nicotinic acid


Formula: C₂₀H₁₂F₅N₃O₄
MW: 453.32 g/mol
Storage Temperature: Freezer
MDL Number: MFCD31813692
CAS Number: 1416799-28-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty