Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AZD-1283 ≥98% (by HPLC), Moligand™

Supplier: ALADDIN SCIENTIFIC

A287112-10MG A287112-50MG A287112-5MG
ALA287112-10MGEA 145.26 USD InStock
ALA287112-10MG ALA287112-50MG ALA287112-5MG
AZD-1283 ≥98% (by HPLC), Moligand™
AZD-1283


Formula: C₂₃H₂₆N₄O₅S
MW: 470.54 g/mol
Storage Temperature: Freezer
MDL Number: MFCD26395344
CAS Number: 919351-41-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty