Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2,5-Dichlorophenyl)-4-(furan-2-yl)-2-methyl-5-oxo-1,4,5,6,7,8-hexahydroquinoline-3-carboxamide ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC

A287232-25MG A287232-50MG A287232-5MG A287232-100MG A287232-10MG A287232-250MG
ALA287232-25MGEA 309.61 USD InStock
ALA287232-25MG ALA287232-50MG ALA287232-5MG ALA287232100MG ALA287232-10MG ALA287232250MG
N-(2,5-Dichlorophenyl)-4-(furan-2-yl)-2-methyl-5-oxo-1,4,5,6,7,8-hexahydroquinoline-3-carboxamide ≥98% (by HPLC)
N-(2,5-Dichlorophenyl)-4-(furan-2-yl)-2-methyl-5-oxo-1,4,5,6,7,8-hexahydroquinoline-3-carboxamide
Product Introduction:AR420626 is a selective agonist of free fatty acid receptor 3 (FFAR3, GPR41) with IC50 of 117 nM. AR420626 inhibits nicotine and serotonin-induced changes in motility of isolated muscle strips from rat colon and suppresses serotonin-induced fecal output in rats。


Formula: C₂₁H₁₈Cl₂N₂O₃
MW: 417.29 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28987470
CAS Number: 1798310-55-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty