Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Bis[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]-1,1,3,3-tetramethyldisiloxane 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A133377-10G A133377-1G A133377-5G
77683-181EA 32 USD InStock
77683-181 77683-182 77683-183
1,3-Bis[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]-1,1,3,3-tetramethyldisiloxane 95%, Ambeed.Inc
1,3-Bis[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]-1,1,3,3-tetramethyldisiloxane
Precaución: For research use only.


Formula: C₂₀H₃₈O₃Si₂
MW: 382.69 g/mol
MDL Number: MFCD03262840
CAS Number: 18724-32-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty