Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1R)-(6-methoxyquinolin-4-yl)((2R,4S,5R)-5-vinylquinuclidin-2-yl)methyl)thiourea 97%, Ambeed.Inc

Supplier: AMBEED, INC

A803063-100MG A803063-250MG
77684-747EA 105 USD InStock
77684-747 77684-748
1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1R)-(6-methoxyquinolin-4-yl)((2R,4S,5R)-5-vinylquinuclidin-2-yl)methyl)thiourea 97%, Ambeed.Inc
1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1R)-(6-methoxyquinolin-4-yl)((2R,4S,5R)-5-vinylquinuclidin-2-yl)methyl)thiourea
Precaución: For research use only.


Formula: C₂₉H₂₈F₆N₄OS
MDL Number: MFCD16621389
CAS Number: 852913-25-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty