96-Well PCR Clean-Up Kits, IBI Scientific
Supplier: Jopak Verpakkingen
IB47040 IB47050
95039-646EA 802.83 USD InStock
95039-646 95039-648
96-Well PCR Clean-Up Kits, IBI Scientific
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The IBI high throughput 96-Well PCR Clean-Up Kit is designed to recover or concentrate DNA fragments from PCR or other enzymatic reactions using an efficient 96-well binding plate system.
  • Sample size: up to 100µl of PCR products per well
  • Fragment size: 70bp – 20Kb
  • Expectant yield: up to 95%
  • Operation time: 20 minutes
Chaotropic salt is used to denature enzymes. DNA fragments in the chaotropic salt are bound by the glass fiber matrix in each well of the binding plate. Contaminants are removed with a Wash Buffer (containing ethanol) and the purified DNA fragments are eluted by a low salt Elution buffer, TE, or water. Salts, enzymes, and unincorporated nucleotides can be effectively removed from the reaction mixture without phenol extraction or alcohol precipitation and the purified DNA is ready for use in subsequent reactions.
The quality of the 96-Well PCR Clean-Up Kit is tested on a lot-to-lot basis by purifying DNA fragments of various sizes from PCR products. The purified DNA is analyzed by electrophoresis.
Caution: Components contain irritant agents. Always wear a lab coat, disposable gloves, and protective goggles during use.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty