Multi-Element Standard 23 components: 50 µg/g each of Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil, VHG, LGC Standards
Supplier: LGC STANDARDS
VHG-V23-50-400G VHG-V23-50-100G VHG-V23-50-200G
77693-314EA 399.9 USD InStock
77693-314 77695-699 77695-700
Multi-Element Standard 23 components: 50 µg/g each of Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty