Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-SAMD8 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG BIOSCIENCE LLC
A61892-50UG A61892-100UG
77719-265EA 228.38 USD InStock
77719-265 77719-264
Anti-SAMD8 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: SAMD8
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty