Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-AGBL1 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A49070-100UL A49070-50UL
77715-756EA 334.95 USD InStock
77715-756 77715-757
Anti-AGBL1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, IHC:1:15-1:50

Type:
Antigen: AGBL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty