Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-At2g24580 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG BIOSCIENCE LLC
A47075-50UG A47075-100UG
77712-333EA 228.38 USD InStock
77712-333 77712-332
Anti-At2g24580 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: At2g24580
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: A. thaliana

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty