Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-KRT6A/KRT6B/KRT6C Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A46977-50UL A46977-100UL
77712-156EA 228.38 USD InStock
77712-156 77712-155
Anti-KRT6A/KRT6B/KRT6C Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, IHC:1:25-1:100

Type:
Antigen: KRT6A/KRT6B/KRT6C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty