Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Serine protease inhibitor Kazal-type 2-like protein Rabbit Polyclonal Antibody (HRP)

Supplier: AFG BIOSCIENCE LLC
A47243-100UG A47243-50UG
77712-616EA 334.95 USD InStock
77712-616 77712-617
Anti-Serine protease inhibitor Kazal-type 2-like protein Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: Serine protease inhibitor Kazal-type 2-like protein
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Cyprinus carpio

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty