Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-PAWR Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A48146-100UL A48146-50UL
77714-157EA 334.95 USD InStock
77714-157 77714-158
Anti-PAWR Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, WB:1:1000-1:3000, IHC:1:25-1:100

Type:
Antigen: PAWR
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty