Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-ACADS Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45119-100UL A45119-50UL
77708-869EA 334.95 USD InStock
77708-869 77708-870
Anti-ACADS Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:25-1:100

Type:
Antigen: ACADS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty