BST Prime DNA Polymerase
Supplier: EMPIRICAL BIOSCIENCE
BST-RB-2000 BST-RB-8000
77676-254EA 75.03 USD InStock
77676-254 77676-256
BST Prime DNA Polymerase
Nucleic Acid Reagents End-point PCR Enzymes and Kits
BST Prime is a recombinantly produced Bacillus stearothermophilus DNA polymerase I, large fragment (BST DNA Polymerase, large fragment).
  • 10X LAMP Reaction storage buffer
  • Applications: Isothermal amplification
BST Prime is a genetically improved polymerase compared to the wild-type enzyme. BST Prime DNA Polymerase contains 5' to 3' DNA polymerase activity and strong strand displacement activity but lacks 5' to 3' exonuclease activity. The enzyme generates an amplification factor of up to 109 which is comparable to approximately 30 cycles in a PCR assay which allows for detection of a target gene in 10 to 30 minutes.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty