Brownlee™ Peco/Fast-LC Cartridge Columns (80 Å Pore Size), PerkinElmer
Supplier: PERKINELMER U.S. LLC
02580191 02580195
97012-344PK 1723.5 USD InStock
97012-344 97012-346 97013-100 97013-114 97013-498 97013-116
Brownlee™ Peco/Fast-LC Cartridge Columns (80 Å Pore Size), PerkinElmer
Chromatography Columns
These cartridge columns are packed with a small-pore, silica-based sorbent for HPLC of small molecules.
  • Fine pore design
  • Silica-based sorbent
  • Suited for HPLC procedures on small molecules
Extremely fine pore size allows for excellent resolving power.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty