032470-M1
032470-K7
AA32470-M1EA
361
USD
InStock
AA32470-M1
AA32470-K2
AA32470-K7
Acetonitrile ≥99.7% for spectrophotometry
Acetonitrile
Purity: 99.7+%
CAS No.: 75-05-8
Molecular Formula: CH3CN
Formula Weight: 41.05
Flash Point: 5°(41°F)
Physical Form: Liquid
Density: 0.786
Refractive Index: 1.3440
Odor: Pungent
Melting Point: -48 to -45°
Boiling Point: 80-82°
Beilstein Reference: 741857
Merck Reference: 14,70
UN No.: 1648
MDL No.: MFCD00001878