Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Silver hard ≥99.95% (metals basis), foil, hard, Thickness: 0.075 mm (0.003 in)

Supplier: Origen Biomedical, Inc

000501-FS 000501-GT
AA00501-FSEA 107 USD InStock
AA00501-FS AA00501-GT
Silver hard ≥99.95% (metals basis), foil, hard, Thickness: 0.075 mm (0.003 in)
Silver
MDL: MFCD00003397

Purity: 99.95% CAS No.: 7440-22-4 Physical Form: ≈3g/25x150mm, Temper: as rolled Odor: Characteristic MDL No.: MFCD00003397


Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty