Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Calcium nitrate hydrate ≥99.995% (metals basis), Puratronic®

Supplier: Origen Biomedical, Inc
Danger

010683-14 010683-22
AA10683-14EA 134 USD InStock
AA10683-14 AA10683-22
Calcium nitrate hydrate ≥99.995% (metals basis), Puratronic®
Calcium nitrate hydrate
MDL: MFCD00149604 Fieser: 18,84

Purity: 99.995% CAS No.: 35054-52-5 Molecular Formula: Ca(NO3)2.xH2O (x≈4) Formula Weight: 164.09(anhy) Physical Form: Crystalline Aggregates Density: 1.82 Melting Point: 42° Boiling Point: 132° dec. Merck Reference: 14,1682 UN No.: 1454 MDL No.: MFCD00149604


Formula: Ca(NO₃)₂·H₂O
MW: 182.1 g/mol
Boiling Pt: 132 °C (dec.)
Melting Pt: 42 °C
Density: 1.82
Storage Temperature: Ambient
MDL Number: MFCD00149604
CAS Number: 35054-52-5
EINECS: 233-332-1
UN: 1454
ADR: 5.1 III
Merck Index: 14,1682
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty